α-Bromo-2,3,4,5-tetrafluorotoluene
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2,3,4,5-Tetrafluorobenzyl bromide
PC0500-25G
PC0500-5G
APOSPC0500-25GEA
454
EUR
APOSPC0500-25G
APOSPC0500-5G
α-Bromo-2,3,4,5-tetrafluorotoluene
α-Bromo-2,3,4,5-tetrafluorotoluene
Formula:
C₇H₃BrF₄ MW: 243 g/mol Boiling Pt: 107…110 °C/65mmHg Storage Temperature: Ambient |
MDL Number:
MFCD01631453 CAS Number: 53001-71-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...