(3-Chloro-5-fluorophenyl)acetic acid

Supplier: Apollo Scientific
Warning

PC0538-1G PC0538-25G PC0538-5G PC0538-100G PC0538-500G
APOSPC0538-1GEA 21.9 EUR
APOSPC0538-1G APOSPC0538-25G APOSPC0538-5G APOSPC0538-100G APOSPC0538-500G
(3-Chloro-5-fluorophenyl)acetic acid
(3-Chloro-5-fluorophenyl)acetic acid
Formula: (C₆H₃ClF)CH₂COOH
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD03788475
CAS Number: 202001-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR