3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene 97%
Supplier: Apollo Scientific
|
Warning
|
PC0553-25G
PC0553-5G
APOSPC0553-25GEA
168
EUR
APOSPC0553-25G
APOSPC0553-5G
3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene 97%
3-NTF-BOA, 5-carboxy-2-(trifluoromethyl)nitrobenzene
Formula:
C₈H₄F₃NO₄ Melting Pt: 169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274359 CAS Number: 116965-16-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...