1-Fluoro-2-iodoethane 98%

Supplier: Apollo Scientific
Danger

PC0560-5G PC0560-1G PC0560-25G
APOSPC0560-5GEA 56.7 EUR
APOSPC0560-5G APOSPC0560-1G APOSPC0560-25G
1-Fluoro-2-iodoethane 98%
1-Fluoro-2-iodoethane
Formula: C₂H₄FI
Boiling Pt: 98…102 °C
Density: 2,136 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00748131
CAS Number: 762-51-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR