(S)-1-(2-Fluorophenyl)ethylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(S)-1-(2-Fluorophenyl)ethanamine
PC0612-1G
PC0612-250MG
APOSPC0612-1GEA
162
EUR
APOSPC0612-1G
APOSPC0612-250MG
(S)-1-(2-Fluorophenyl)ethylamine
(S)-1-(2-Fluorophenyl)ethylamine
Formula:
C₈H₁₀FN MW: 139,17 g/mol Boiling Pt: 179,8 °C Density: 1,063 g/cm³ (25 °C) Flash Pt: 70,2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03092994 CAS Number: 68285-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...