Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

PC0654-1G PC0654-5G
APOSPC0654-1GEA 109 EUR
APOSPC0654-1G APOSPC0654-5G
Ethyl-4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate
Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate
Formula: C₁₃H₁₀F₃NO₃
MW: 285,2200 g/mol
Melting Pt: 336 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00173369
CAS Number: 26893-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR