4-Fluoro-D-phenylalanine

Supplier: Apollo Scientific

Synonyms: (R)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-D-Phe(4-F)-OH

PC0660-5G
APOSPC0660-5GEA 66.7 EUR
APOSPC0660-5G
4-Fluoro-D-phenylalanine
4-Fluoro-D-phenylalanine
Formula: C₉H₁₀FNO₂
MW: 183,1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063063
CAS Number: 18125-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR