1H,1H,5H-Octafluoropentyl-p-toluenesulphonate 96%
Supplier: Apollo Scientific
PC0772-25G
PC0772-5G
PC0772-100G
APOSPC0772-25GEA
205
EUR
APOSPC0772-25G
APOSPC0772-5G
APOSPC0772-100G
1H,1H,5H-Octafluoropentyl-p-toluenesulphonate 96%
1H,1H,5H-Octafluoropentyl-p-toluenesulphonate
Formula:
C₁₂H₁₀F₈O₃S MW: 386,26 g/mol Boiling Pt: 157 °C/5mmHg Melting Pt: 44538 °C Density: 1,5127 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00378586 CAS Number: 2264-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...