3-Bromophenylsulphur pentafluoride 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-3-(pentafluorosulphanyl)benzene

PC0782-1G PC0782-25G PC0782-5G
APOSPC0782-1GEA 285 EUR
APOSPC0782-1G APOSPC0782-25G APOSPC0782-5G
3-Bromophenylsulphur pentafluoride 95%
3-Bromophenylsulphur pentafluoride
Formula: C₆H₄BrF₅S
MW: 283,06 g/mol
Density: 1,831 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD03425927
CAS Number: 672-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR