2-Fluoro-4-(trifluoromethyl)cinnamic acid 98%

Supplier: Apollo Scientific

Synonyms: 3-(2-Fluoro-4-(trifluoromethyl)phenyl)acrylic acid

PC0783-10G PC0783-5G PC0783-1G
APOSPC0783-10GEA 186 EUR
APOSPC0783-10G APOSPC0783-5G APOSPC0783-1G
2-Fluoro-4-(trifluoromethyl)cinnamic acid 98%
2-Fluoro-4-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00236292
CAS Number: 262608-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR