4-(Pentafluorothio)phenol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Hydroxyphenylsulphur pentafluoride

PC0794-1G PC0794-5G
APOSPC0794-1GEA 551 EUR
APOSPC0794-1G APOSPC0794-5G
4-(Pentafluorothio)phenol 97%
4-(Pentafluorothio)phenol
Formula: C₆H₅F₅OS
MW: 220,16 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD03788516
CAS Number: 774-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR