3-(Pentafluorothio)benzoic acid 95%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Carboxyphenylsulphur pentafluoride, 3-Sulphurpentafluorobenzoic acid
PC0800-1G
PC0800-5G
APOSPC0800-1GEA
430
EUR
APOSPC0800-1G
APOSPC0800-5G
3-(Pentafluorothio)benzoic acid 95%
3-(Pentafluorothio)benzoic acid
Formula:
C₇H₅F₅O₂S MW: 248,17 g/mol Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788519 CAS Number: 833-96-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...