3-(Pentafluorosulphanyl)benzoyl chloride
Supplier: Apollo Scientific
PC0807-5G
PC0807-1G
APOSPC0807-5GEA
1020
EUR
APOSPC0807-5G
APOSPC0807-1G
3-(Pentafluorosulphanyl)benzoyl chloride
3-(Pentafluorosulphanyl)benzoyl chloride
Formula:
C₇H₄ClF₅OS MW: 266.62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788522 CAS Number: 401892-81-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...