2-Fluoro-6-methoxyaniline
Supplier: Apollo Scientific
|
Warning
|
PC0872-1G
PC0872-250MG
PC0872-5G
APOSPC0872-1GEA
24.8
EUR
APOSPC0872-1G
APOSPC0872-250MG
APOSPC0872-5G
2-Fluoro-6-methoxyaniline
2-Fluoro-6-methoxyaniline
Formula:
C₇H₈FNO Boiling Pt: 208 °C/756mmHg Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788538 CAS Number: 446-61-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...