1,2-Dibromo-1-chloro-1,2,2-trifluoroethane 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloro-1,2-dibromo-1,1,2-trifluoroethane, 1-Chloro-1,2-dibromo-1,2,2-trifluoroethane

PC0898-25G PC0898-100G PC0898-500G
APOSPC0898-25GEA 55.3 EUR
APOSPC0898-25G APOSPC0898-100G APOSPC0898-500G
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane 98%
1,2-Dibromo-1-chloro-1,2,2-trifluoroethane
Formula: BrClFCCBrF₂
MW: 276,28 g/mol
Boiling Pt: 93…94 °C
Melting Pt: –72,9 °C
Density: 2,248 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039316
CAS Number: 354-51-8
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR