(2-Fluoro-6-methoxyphenyl)acetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC0906-10G
PC0906-1G
PC0906-5G
APOSPC0906-10GEA
238
EUR
APOSPC0906-10G
APOSPC0906-1G
APOSPC0906-5G
(2-Fluoro-6-methoxyphenyl)acetic acid 98%
(2-Fluoro-6-methoxyphenyl)acetic acid
Formula:
C₉H₉FO₃ MW: 184,16 g/mol Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671770 CAS Number: 500912-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...