Tropylium tetrafluoroborate 95%

Supplier: Apollo Scientific
Danger

Synonyms: Tropyliumtetrafluoroborate, Cyclohepta-2,4,6-trienylium tetrafluoroborate

PC0915-25G PC0915-5G
APOSPC0915-25GEA 255 EUR
APOSPC0915-25G APOSPC0915-5G
Tropylium tetrafluoroborate 95%
Tropylium tetrafluoroborate
Formula: C₇H₇BF₄
MW: 177.94 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013402
CAS Number: 27081-10-3
EINECS: 248-214-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR