3-[6-(Trifluoromethyl)pyridin-3-yl]propanoic acid 95+%

Supplier: Apollo Scientific
Warning

PC0983-10G PC0983-1G PC0983-5G
APOSPC0983-10GEA 2320 EUR
APOSPC0983-10G APOSPC0983-1G APOSPC0983-5G
3-[6-(Trifluoromethyl)pyridin-3-yl]propanoic acid 95+%
3-[6-(Trifluoromethyl)pyridin-3-yl]propanoic acid
Formula: C₉H₈F₃NO₂
MW: 219,16 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD06801326
CAS Number: 539855-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR