Aluminium fluoride, anhydrous 99%

Supplier: Apollo Scientific
Warning

Synonyms: Aluminum fluoride

PC1008-100G PC1008-25G
APOSPC1008-100GEA 234 EUR
APOSPC1008-100G APOSPC1008-25G
Aluminium fluoride, anhydrous 99%
Aluminium fluoride
Formula: AlF₃
MW: 83,98 g/mol
Melting Pt: 1290 °C (subl.)
Density: 3,1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003426
CAS Number: 7784-18-1
EINECS: 232-051-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR