3,5-Dimethyl-2,4,6-trifluorobenzotrifluoride
Supplier: Apollo Scientific
|
Warning
|
PC10083-1G
PC10083-5G
APOSPC10083-1GEA
223
EUR
APOSPC10083-1G
APOSPC10083-5G
3,5-Dimethyl-2,4,6-trifluorobenzotrifluoride
3,5-Dimethyl-2,4,6-trifluorobenzotrifluoride
Formula:
C₉H₆F₆ Storage Temperature: Ambient |
CAS Number:
886762-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...