2,4,6-Trifluorotoluene 95%
Supplier: Apollo Scientific
|
Danger
|
PC10093-5G
PC10093-1G
APOSPC10093-5GEA
560
EUR
APOSPC10093-5G
APOSPC10093-1G
2,4,6-Trifluorotoluene 95%
2,4,6-Trifluorotoluene
Formula:
C₇H₅F₃ Boiling Pt: 105…110 °C Density: 1.23 g/cm³ (25 °C) Flash Pt: 20 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039204 CAS Number: 93343-11-4 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...