5-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-indole-2,3-dione

Supplier: Apollo Scientific

PC10134-1G PC10134-500MG PC10134-5G
APOSPC10134-1GEA 217 EUR
APOSPC10134-1G APOSPC10134-500MG APOSPC10134-5G
5-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-indole-2,3-dione
5-Chloro-1-[3-(trifluoromethyl)benzyl]-1H-indole-2,3-dione
Formula: C₁₆H₉ClF₃NO₂
Melting Pt: 165,5…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00141725
CAS Number: 89159-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR