3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid

Supplier: Apollo Scientific

PC10189-1G PC10189-500MG PC10189-5G
APOSPC10189-1GEA 405 EUR
APOSPC10189-1G APOSPC10189-500MG APOSPC10189-5G
3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid
3-[2-(4-Bromo-2-fluorophenoxy)phenyl]acrylic acid
Formula: C₁₅H₁₀BrFO₃
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD01814603
CAS Number: 449778-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR