1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone

Supplier: Apollo Scientific

Synonyms: 2-Acetyl-5-(4-fluorophenyl)thiophene

PC10202-1G PC10202-500MG PC10202-5G
APOSPC10202-1GEA 263 EUR
APOSPC10202-1G APOSPC10202-500MG APOSPC10202-5G
1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone
1-[5-(4-Fluorophenyl)-2-thienyl]-1-ethanone
Formula: C₁₂H₉FOS
MW: 220,27 g/mol
Melting Pt: 119…120 °C
Storage Temperature: Ambient
MDL Number: MFCD06200932
CAS Number: 886361-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR