Ethyl-2-{1-[3-(trifluoromethyl)benzoyl]-4-piperidinylidene}acetate

Supplier: Apollo Scientific

PC10219-1G PC10219-500MG
APOSPC10219-1GEA 402 EUR
APOSPC10219-1G APOSPC10219-500MG
Ethyl-2-{1-[3-(trifluoromethyl)benzoyl]-4-piperidinylidene}acetate
Ethyl-2-{1-[3-(trifluoromethyl)benzoyl]-4-piperidinylidene}acetate
Formula: C₁₇H₁₈F₃NO₃
MW: 341,33 g/mol
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD03425754
CAS Number: 439095-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR