4-Fluorobenzo[b]thiophene-2-carboxylic acid

Supplier: Apollo Scientific
Warning

PC10244-1G PC10244-250MG
APOSPC10244-1GEA 98.9 EUR
APOSPC10244-1G APOSPC10244-250MG
4-Fluorobenzo[b]thiophene-2-carboxylic acid
4-Fluorobenzo[b]thiophene-2-carboxylic acid
Formula: C₉H₅FO₂S
MW: 196,1982 g/mol
Melting Pt: 217…220 °C
Storage Temperature: Ambient
MDL Number: MFCD03425782
CAS Number: 310466-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR