1-(4-Amino-2-fluorophenyl)-4-methylpiperazine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-4-(4-methylpiperazino)aniline, 3-fluoro-4-(4-methyl-1-piperazinyl)benzenamine, 3-Fluoro-4-(4-methylpiperazin-1-yl)aniline

PC10264-10G PC10264-1G
APOSPC10264-10GEA 118 EUR
APOSPC10264-10G APOSPC10264-1G
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
1-(4-Amino-2-fluorophenyl)-4-methylpiperazine
Formula: C₁₁H₁₆FN₃
MW: 209,27 g/mol
Melting Pt: 87…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663793
CAS Number: 221198-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR