1,1,1-Trifluoro-3-(phenethylamino)-2-propanol
Supplier: Apollo Scientific
Synonyms:
3-Phenethylamino-1,1,1-trifluoropropan-2-ol
PC10271-1G
PC10271-500MG
APOSPC10271-1GEA
799
EUR
APOSPC10271-1G
APOSPC10271-500MG
1,1,1-Trifluoro-3-(phenethylamino)-2-propanol
1,1,1-Trifluoro-3-(phenethylamino)-2-propanol
Formula:
C₁₁H₁₄F₃NO Melting Pt: 63…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD03001278 CAS Number: 400878-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...