Pentafluorophenyl-5-chloro-2-methoxybenzenesulphonate

Supplier: Apollo Scientific

PC10304-1G PC10304-5G
APOSPC10304-1GEA 201 EUR
APOSPC10304-1G APOSPC10304-5G
Pentafluorophenyl-5-chloro-2-methoxybenzenesulphonate
Pentafluorophenyl 5-chloro-2-methoxybenzenesulphonate
Formula: C₁₃H₆ClF₅O₄S
Melting Pt: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD05975152
CAS Number: 886361-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR