α,α,α-Trifluoro-m-toluidine 98%

Supplier: Apollo Scientific
Danger

Synonyms: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride

PC1030A-1KG PC1030A-250G
APOSPC1030A-1KGEA 94.4 EUR
APOSPC1030A-1KG APOSPC1030A-250G
α,α,α-Trifluoro-m-toluidine 98%
α,α,α-Trifluoro-m-toluidine

Intermediate for the synthesis of several pharmaceuticals including Bendrofluazide

Formula: C₇H₆F₃N
MW: 161,13 g/mol
Boiling Pt: 187 °C
Melting Pt: 44352 °C
Density: 1,29 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007797
CAS Number: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR