2-{[(4-Fluorophenyl)sulphonyl]amino}acetic acid

Supplier: Apollo Scientific

PC10351-1G PC10351-250MG
APOSPC10351-1GEA 127 EUR
APOSPC10351-1G APOSPC10351-250MG
2-{[(4-Fluorophenyl)sulphonyl]amino}acetic acid
2-{[(4-Fluorophenyl)sulphonyl]amino}acetic acid
Formula: C₈H₈FNO₄S
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD02091617
CAS Number: 13029-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR