Methyl-3,4,5-trifluorobenzenecarboxylate 99%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 3,4,5-trifluorobenzoate

PC10372-1G PC10372-25G PC10372-5G PC10372-100G
APOSPC10372-1GEA 14.7 EUR
APOSPC10372-1G APOSPC10372-25G APOSPC10372-5G APOSPC10372-100G
Methyl-3,4,5-trifluorobenzenecarboxylate 99%
Methyl-3,4,5-trifluorobenzenecarboxylate
Formula: C₈H₅F₃O₂
MW: 190,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200977
CAS Number: 773873-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR