2-Bromo-2'-chloro-6'-fluoroacetophenone 98%

Supplier: Apollo Scientific
Danger

PC10428-1G PC10428-5G PC10428-25G
APOSPC10428-1GEA 62.5 EUR
APOSPC10428-1G APOSPC10428-5G APOSPC10428-25G
2-Bromo-2'-chloro-6'-fluoroacetophenone 98%
2-Bromo-2'-chloro-6'-fluoroacetophenone
Formula: C₈H₅BrClFO
Boiling Pt: 74 °C/0,06mmHg
Storage Temperature: Ambient
MDL Number: MFCD07368692
CAS Number: 157359-99-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR