3-Nitro-4-fluorobenzoyl chloride

Supplier: Apollo Scientific
Danger

Synonyms: 4-Fluoro-3-nitrobenzoyl chloride

PC10445-1G PC10445-5G
APOSPC10445-1GEA 21.9 EUR
APOSPC10445-1G APOSPC10445-5G
3-Nitro-4-fluorobenzoyl chloride
3-Nitro-4-fluorobenzoyl chloride
Formula: C₇H₃ClFNO₃
Boiling Pt: 153 °C/12mmHg
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD07368701
CAS Number: 400-94-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR