2-Fluoronicotinonitrile 98%

Supplier: Apollo Scientific
Warning

PC10522-100G PC10522-25G PC10522-5G
APOSPC10522-100GEA 116 EUR
APOSPC10522-100G APOSPC10522-25G APOSPC10522-5G
2-Fluoronicotinonitrile 98%
2-Fluoronicotinonitrile
Formula: C₆H₃FN₂
Melting Pt: 27…33 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD03095082
CAS Number: 3939-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR