ω,ω-Dibromo-4-fluoroacetophenone

Supplier: Apollo Scientific
Danger

PC10523-1G PC10523-5G
APOSPC10523-1GEA 93.1 EUR
APOSPC10523-1G APOSPC10523-5G
ω,ω-Dibromo-4-fluoroacetophenone
ω,ω-Dibromo-4-fluoroacetophenone
Formula: C₈H₅Br₂FO
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD03791309
CAS Number: 7542-64-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR