2-Fluoro-3-pyridineboronic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Fluoro-3-pyridinylboronic acid, (2-Fluoropyridin-3-yl)boronic acid, 2-Fluoropyridine-3-boronic acid
PC10537-25G
PC10537-5G
APOSPC10537-25GEA
42.2
EUR
APOSPC10537-25G
APOSPC10537-5G
2-Fluoro-3-pyridineboronic acid
2-Fluoro-3-pyridineboronic acid
Formula:
C₅H₅BFNO₂ Melting Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411557 CAS Number: 174669-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...