2,2-Difluorobutyric acid 97%

Supplier: Apollo Scientific
Danger

PC1055-1G PC1055-5G
APOSPC1055-1GEA 129 EUR
APOSPC1055-1G APOSPC1055-5G
2,2-Difluorobutyric acid 97%
2,2-Difluorobutyric acid
Formula: C₄H₆F₂O₂
MW: 124,09 g/mol
Boiling Pt: 100…101 °C/8,5mmHg
Melting Pt: 44446 °C
Density: 1,215 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD03093765
CAS Number: 2366-62-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR