2-(2-Fluorophenyl)quinoline-4-carboxylic acid

Supplier: Apollo Scientific
Warning

PC10563-1G PC10563-5G
APOSPC10563-1GEA 147 EUR
APOSPC10563-1G APOSPC10563-5G
2-(2-Fluorophenyl)quinoline-4-carboxylic acid
2-(2-Fluorophenyl)quinoline-4-carboxylic acid
Formula: C₁₆H₁₀FNO₂
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD06659551
CAS Number: 1647-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR