2-(2,4-Difluoroanilino)acetohydrazide 95%

Supplier: Apollo Scientific
Warning

Synonyms: N-(2,4-Difluorophenyl)glycinehydrazide

PC10589-1G PC10589-250MG
APOSPC10589-1GEA 90.2 EUR
APOSPC10589-1G APOSPC10589-250MG
2-(2,4-Difluoroanilino)acetohydrazide 95%
2-(2,4-Difluoroanilino)acetohydrazide
Formula: C₈H₉F₂N₃O
MW: 201,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00098762
CAS Number: 2351-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR