3,3,3-Trifluoropropionyl chloride
Supplier: Apollo Scientific
|
Danger
|
PC10626-10G
PC10626-25G
PC10626-5G
APOSPC10626-10GEA
143
EUR
APOSPC10626-10G
APOSPC10626-25G
APOSPC10626-5G
3,3,3-Trifluoropropionyl chloride
3,3,3-Trifluoropropionyl chloride
Formula:
C₃H₂ClF₃O Boiling Pt: 71…72 °C Density: 1,422 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00800953 CAS Number: 41463-83-6 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...