Tributylphosphonium tetrafluoroborate

Supplier: Apollo Scientific
Warning

Synonyms: Tri-n-butylphosphonium tetrafluoroborate

PC10682-1G PC10682-25G PC10682-5G
APOSPC10682-1GEA 21.9 EUR
APOSPC10682-1G APOSPC10682-25G APOSPC10682-5G
Tributylphosphonium tetrafluoroborate
Tributylphosphonium tetrafluoroborate
Formula: C₁₂H₂₈BF₄P
MW: 290,13 g/mol
Melting Pt: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD05664257
CAS Number: 113978-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR