Difluoromethyl 2-Pyridyl sulphone 97%

Supplier: Apollo Scientific

Synonyms: 2-((Difluoromethyl)sulphonyl)pyridine, 2-PySO2CF2H, 2-(Difluoromethylsulphonyl)pyridine

PC10687-25G
APOSPC10687-25GEA 112 EUR
APOSPC10687-25G
Difluoromethyl 2-Pyridyl sulphone 97%
Difluoromethyl 2-Pyridyl sulphone
Formula: C₆H₅F₂NO₂S
MW: 193,17 g/mol
Melting Pt: 44…49 °C
Storage Temperature: Ambient
MDL Number: MFCD17010193
CAS Number: 1219454-89-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR