1,1,1-Trifluoropropan-2-amine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
1-(Trifluoromethyl)ethylamine, 2-Amino-1,1,1-trifluoropropane
PC10716-1G
APOSPC10716-1GEA
34.8
EUR
APOSPC10716-1G
1,1,1-Trifluoropropan-2-amine 97%
1,1,1-Trifluoropropan-2-amine
Formula:
C₃H₆F₃N MW: 113,08 g/mol Boiling Pt: 46…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973020 CAS Number: 421-49-8 UN: 2733 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...