2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-4-hydroxy-6-trifluoromethylpyrimidine

PC1076Y-1G PC1076Y-25G PC1076Y-5G
APOSPC1076Y-1GEA 14.7 EUR
APOSPC1076Y-1G APOSPC1076Y-25G APOSPC1076Y-5G
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine 97%
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine
Formula: C₅H₄F₃N₃O
MW: 179,1 g/mol
Melting Pt: 288…292 °C
Storage Temperature: Ambient
MDL Number: MFCD00068093
CAS Number: 1513-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR