3-Amino-4-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
|
Warning
|
PC1100I-1G
PC1100I-5G
PC1100I-10G
APOSPC1100I-1GEA
58.2
EUR
APOSPC1100I-1G
APOSPC1100I-5G
APOSPC1100I-10G
3-Amino-4-(trifluoromethyl)pyridine 97%
3-Amino-4-(trifluoromethyl)pyridine
Formula:
C₆H₅F₃N₂ Boiling Pt: 63 °C/1,5mmHg Melting Pt: 55…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173960 CAS Number: 175204-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...