2-Amino-3,6-difluorobenzonitrile
Supplier: Apollo Scientific
PC110110-1G
PC110110-5G
APOSPC110110-1GEA
184
EUR
APOSPC110110-1G
APOSPC110110-5G
2-Amino-3,6-difluorobenzonitrile
2-Amino-3,6-difluorobenzonitrile
Formula:
C₇H₄F₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD13178244 CAS Number: 190011-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...