4-Chloro-3-(trifluoromethyl)phenylboronic acid 95%

Supplier: Apollo Scientific
Warning

PC11111-25G PC11111-5G PC11111-1G
APOSPC11111-25GEA 267 EUR
APOSPC11111-25G APOSPC11111-5G APOSPC11111-1G
4-Chloro-3-(trifluoromethyl)phenylboronic acid 95%
4-Chloro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD03094999
CAS Number: 176976-42-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR