Ammonium trifluoroacetate 98%

Supplier: Apollo Scientific
Warning

Synonyms: Ammoniumtrifluoroacetate, Trifluoroacetic acid ammonium salt

PC1113-100G
APOSPC1113-100GEA 144 EUR
APOSPC1113-100G
Ammonium trifluoroacetate 98%
Ammonium Trifluoroacetate

Mass spec standard for HPLC/MS, Org. Mass. Spec., 187, 25, 1990

Formula: C₂H₄F₃NO₂
MW: 131,05 g/mol
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00012615
CAS Number: 3336-58-1
EINECS: 222-072-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR