4'-Fluorobiphenyl-3-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

PC11143-5G
APOSPC11143-5GEA 142 EUR
APOSPC11143-5G
4'-Fluorobiphenyl-3-carboxylic acid 95%
4'-Fluorobiphenyl-3-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00452724
CAS Number: 10540-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR